Order Entry
China
ContactUsLinkComponent
Tetrabutylammonium hydroxide 40% in aqueous solution, Sigma-Aldrich®
Corrosive
Tetrabutylammonium hydroxide 40% in aqueous solution, Sigma-Aldrich®
Catalog # SIAL86880-500ML
Supplier:  SIGMAALDRICH
CAS Number:  
Corrosive
Tetrabutylammonium hydroxide 40% in aqueous solution, Sigma-Aldrich®
Catalog # SIAL86880-500ML
Supplier:  SIGMAALDRICH
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    500 mL
  • Seq:
    2725

Specifications

About this item

 :
 :259.48 g/mol
ADR:8,II
 :M-Clarity™ quality level = MQ200
EINECS:218-147-6
UN:3267
 :GHS05
Storage Temperature:Refrigerator
CAS Number:2052-49-5
Formula:C₁₆H₃₇NO
MDL Number:MFCD00009425