Order Entry
China
ContactUsLinkComponent
Ammonium iron(II) sulphate 0.12 mol/l (0.12 N) in aqueous solution, AVS TITRINORM volumetric solution, for COD determination according to NF T 90-101 standard
Ammonium iron(II) sulphate 0.12 mol/l (0.12 N) in aqueous solution, AVS TITRINORM volumetric solution, for COD determination according to NF T 90-101 standard
Catalog # VWRC30884.291
Supplier:  VWR Chemicals
CAS Number:  
undefined
Ammonium iron(II) sulphate 0.12 mol/l (0.12 N) in aqueous solution, AVS TITRINORM volumetric solution, for COD determination according to NF T 90-101 standard
Catalog # VWRC30884.291
Supplier:  VWR Chemicals
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    1 L
  • Seq:
    5037

Specifications

Specification Test Results

  • Titer (20°C, at filling):
    0.1198 - 0.1202 mol/l
  • NIST traceable:
    Confirmed

Specification Test Results

About this item

 :
 :284.05 g/mol
Storage Temperature:Ambient
EINECS:233-151-8
CAS Number:10045-89-3
Formula:H₈FeN₂O₈S₂
MDL Number:MFCD00051039