Order Entry
China
ContactUsLinkComponent
 
Tetrabutylammonium hydroxide 1 mol/l (1 N) in methanol, Sigma-Aldrich®
Catalog # SIAL230189-100ML
Supplier:  SIGMAALDRICH
CAS Number:  
Tetrabutylammonium hydroxide 1 mol/l (1 N) in methanol, Sigma-Aldrich®
Catalog # SIAL230189-100ML
Supplier:  SIGMAALDRICH
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    100 mL
  • Seq:
    2440

Specifications

About this item

 :
 :259.48 g/mol
ADR:3,II
 :M-Clarity™ quality level = MQ200
Storage Temperature:Refrigerator
EINECS:218-147-6
UN:2924
CAS Number:2052-49-5
Formula:C₁₆H₃₇NO
MDL Number:MFCD00009425