Order Entry
China
ContactUsLinkComponent
Magnesium chloride 1 mol/l (2 N) in aqueous solution, Sigma-Aldrich®
Magnesium chloride 1 mol/l (2 N) in aqueous solution, Sigma-Aldrich®
Catalog # SIALM1028-10X1ML
Supplier:  SIGMAALDRICH
CAS Number:  
Magnesium chloride 1 mol/l (2 N) in aqueous solution, Sigma-Aldrich®
Catalog # SIALM1028-10X1ML
Supplier:  SIGMAALDRICH
CAS Number:  

Specifications

  • Pk:
    1
  • Seq:
    2076
  • Pk Info:
    10×1 ml

Specifications

About this item

 :
 :95.21 g/mol
Merck Index:13,05684
 :M-Clarity™ quality level = MQ300
Storage Temperature:Ambient
EINECS:232-094-6
CAS Number:7786-30-3
Formula:MgCl₂
MDL Number:MFCD00011106