
Sodium hydroxide 1 mol/l (1 N) in aqueous solution, Sigma-Aldrich®
SIALS2770-100ML
:
- Pack type:Glass bottle
- Pk:100 mL
- Seq:2440
:
:40 g/mol
:1.04 g/cm³ (25 °C)
:Ambient
:3266
:8,III
:M-Clarity™ quality level = MQ200
:215-185-5
:GHS05
:1310-73-2
:NaOH
:MFCD00003548