Order Entry
China
ContactUsLinkComponent
EDTA disodium salt 0.1 M EDTA-Na2 in aqueous solution Reag. Ph. Eur. volumetric solution, for complexometry, Fluka™
EDTA disodium salt 0.1 M EDTA-Na2 in aqueous solution Reag. Ph. Eur. volumetric solution, for complexometry, Fluka™
  HONC34550-500ML
 :  HONEYWELL
 :  
EDTA disodium salt 0.1 M EDTA-Na2 in aqueous solution Reag. Ph. Eur. volumetric solution, for complexometry, Fluka™
  HONC34550-500ML
 :  HONEYWELL
 :  
  • Pack type:
    Plastic bottle
  • Pk:
    500 mL
  • Seq:
    2725
  • Factor:
    0.999 - 1.001
  • Traceable to NIST SRM:
    effective
 :
 :336.21 g/mol
 :1.01 g/cm³
 :Ambient
 :382-266-9
 :139-33-3
 :C₁₀H₁₄N₂Na₂O₈
 :MFCD00070672