Order Entry
China
Orders LinkContactUsLinkComponent
(±)-1-Ethoxy-2-propanol 95%

(±)-1-Ethoxy-2-propanol 95%

Irritant
Flammable
(±)-1-Ethoxy-2-propanol 95%
Certificates
(±)-1-Ethoxy-2-propanol 95%

2 Options Available Below

Product Details & Documents
Synonyms:  1-Ethoxy-2-propanol, (±)-Monopropylene glycol ethyl ether
  • CAS Number:1569-02-4
  • Formula:C₅H₁₂O₂
  • MDL Number:MFCD00067050
  • Molecular Weight:104.15 g/mol
  • Boiling Pt:132 °C (760 mmHg)
  • Melting Pt:–100 °C
  • Density:0.897 g/cm³
  • Flash Pt:42 °C
  • Storage Temperature:Ambient
  • EINECS:216-374-5
  • UN:3271
  • ADR:3,III

Documents

Specifications for Product Family

Specification Test Results

  • Appearance:
    Clear colorless Liquid
  • Infrared spectrum:
    Conforms
  • Refractive index:
    1.4050 to 1.4090 (20°C, 589 nm)
  • Water:
    ≤0.1 % (K.F.)
  • GC:
    ≥94 % Remark: balance 2-ethoxy-1-propanol
Product Family Options

Product Information