Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
¥0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
China
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
(±)-1-Aminopropan-2-ol
(±)-1-Aminopropan-2-ol 94% contains approx. 5% 2-Amino-1-propanol
(±)-1-Aminopropan-2-ol 94% contains approx. 5% 2-Amino-1-propanol
Specifications
Environmentally Preferable:
Specifications
Specification Test Results
Appearance:
Clear liquid
Infrared spectrum:
Conforms
Refractive index:
1.4470 to 1.4490 (20°C, 589 nm)
Water:
≤0.5 % (K.F.)
GC:
≥93 %
Color scale:
≤50 APHA
Specification Test Results
Related Information
Documentation
SDS
ACRO14965
Documentation