

Chloroform ≥99.8% stabilised, HiPerSolv CHROMANORM® for HPLC
Supplier: VWR Chemicals
Certificates
About this item
CAS Number:67-66-3
:
Formula:CHCl₃
MDL Number:MFCD00000826
:119.38 g/mol
Merck Index:13,02160
Boiling Pt:61.7 °C (1013 hPa)
Melting Pt:–63 °C
Density:1.48320 g/cm³ (20 °C)
EINECS:200-663-8
UN:1888
ADR:6.1,III
:Filtered through a 0.2 µm filter, packaged under nitrogen.
:Stabilised with ethanol 0.6 %
REACH:01-2119486657-20
:GHS08
:GHS06
Storage Temperature:Ambient
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specification Test Results
- Assay (+ stabilizing agent):Min. 99.8 %
- Acidity:Max. 0.0005 meq/g
- Alkalinity:Max. 0.0002 meq/g
- Evaporation residue:Max. 10 ppm
- Water:Max. 0.05 %
- Absorbance (250 nm):Max. 0.3
- Absorbance (260 nm):Max. 0.1
- Absorbance (280 nm):Max. 0.02
- Absorbance (300 nm):Max. 0.01
- Transmittance (250 nm):Min. 50 %
- Transmittance (260 nm):Min. 84 %
- Transmittance (280 nm):Min. 95 %
- Transmittance (300 nm):Min. 98 %
Specification Test Results
Related Information
Certificates
Search by Lot Number1L Glass bottle for liquids DIN 45 (ZPACK13415101) 2,5L Glass bottle for liquids DIN 45 (ZPACK13415249)
Packaging Information
spec_VWRC83627.290 spec_VWRC83627.290 spec_VWRC83627.320 spec_VWRC83627.320