Order Entry
China
ContactUsLinkComponent
di-Sodium L(+)-tartrate dihydrate, Sigma-Aldrich®
di-Sodium L(+)-tartrate dihydrate, Sigma-Aldrich®
Catalog # SIALS4797-100G
Supplier:  SIGMAALDRICH
CAS Number:  
undefined
di-Sodium L(+)-tartrate dihydrate, Sigma-Aldrich®
Catalog # SIALS4797-100G
Supplier:  SIGMAALDRICH
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    100 g
  • Seq:
    3401

Specifications

About this item

 :
 :230.08 g/mol
Merck Index:12,08833
Melting Pt:57 °C
Density:1.818 g/cm³ (20 °C)
 :M-Clarity™ quality level = MQ200
EINECS:212-773-3
CAS Number:6106-24-7
Formula:NaOOC(CHOH)₂COONa·2H₂O
MDL Number:MFCD00150035