Order Entry
China
ContactUsLinkComponent
Tween® 20 (Polysorbate) 100% Nonionic Detergent, Enzyme Immunoassay Grade
Tween® 20 (Polysorbate) 100% Nonionic Detergent, Enzyme Immunoassay Grade
Catalog # BRAD170-6531
Supplier:  BIO RAD
CAS Number:  
Tween® 20 (Polysorbate) 100% Nonionic Detergent, Enzyme Immunoassay Grade
Catalog # BRAD170-6531
Supplier:  BIO RAD
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    100 mL
  • Seq:
    2440
  • Environmentally Preferable:

Specifications

About this item

Synonyms: Polyoxyethylene sorbitan monolaurate
 :
 :1227.53 g/mol
Density:1.1 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:500-018-3
CAS Number:9005-64-5
Formula:C₅₈H₁₁₄O₂₆
MDL Number:MFCD00165986