Polysorbate 20
Catalog # ALFAL15029.AP
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:500 mL
- Seq:2725
Specifications
About this item
:
Boiling Pt:>100 °C
Density:1.105
Flash Pt:>110 °C (230 °F)
Storage Temperature:Ambient
EINECS:500-018-3
CAS Number:9005-64-5
Formula:C₅₈H₁₁₄O₂₆
MDL Number:MFCD00165986