Order Entry
China
ContactUsLinkComponent
TRITON™ X-100 (Polyethylene glycol tert-octylphenyl ether), Reagent Grade for biochemistry
Irritant
Corrosive
Environment
TRITON™ X-100 (Polyethylene glycol tert-octylphenyl ether), Reagent Grade for biochemistry
Supplier:  VWR Chemicals
Certificates
undefined
Irritant
Corrosive
Environment
TRITON™ X-100 (Polyethylene glycol tert-octylphenyl ether), Reagent Grade for biochemistry
Supplier:  VWR Chemicals
Certificates

Specification Test Results

  • pH (5%, Water) @25 °C:
    6 – 8
  • Color (APHA):
    ≤100

Specification Test Results

Related Information