Polyethylene glycol 200 for synthesis, Sigma-Aldrich®
Catalog # MERC8.07483.1000
Supplier: MERCK MILLIPORE
CAS Number:
Specifications
- Pack type:Plastic bottle
- Pk:1 L
- Seq:5037
Specifications
Specification Test Results
- Solubility:(20 °C) soluble
- Melting Point:-38 - -36 °C
- Molar Mass:190 - 210 g/mol
- Boiling Point:150 °C
- Vapor Pressure:- 0.01 hPa (20 °C)
- Flash Point:180 °C
- Density:1.124 g/cm3 (20 °C)
- pH:5 - 7 (100 g/l, H2O, 20 °C)
- Ignition Point:304 °C
- Viscosity kinematic:53 - 62 mm2/s (20 °C)
Specification Test Results
About this item
:
:200 g/mol
Boiling Pt:∼250 °C (1013 hPa)
Melting Pt:–55…–40 °C
Density:1.124 g/cm³ (20 °C)
:M-Clarity™ quality level = MQ200
Flash Pt:180 °C
Storage Temperature:Ambient
EINECS:500-038-2
CAS Number:25322-68-3
:MFCD00081839
Formula:H(OCH₂CH₂)nOH
MDL Number:MFCD01779596