Order Entry
China
ContactUsLinkComponent
 
 
 
 
Benzalkonium chloride 50% (w/w) in aqueous solution
  ALFAB20760.0F
 :  
 
 
 
Benzalkonium chloride 50% (w/w) in aqueous solution
  ALFAB20760.0F
 :  

 

  • Pack type:
    Glass bottle
  • Pk:
    2.500 mL
  • Seq:
    2895

 

 

 :
 :14,1059
 :0.99
 :Ambient
 :264-151-6
 :3265
 :8,III
 :GHS05
 :GHS07
 :GHS09
 :63449-41-2
 :8001-54-5
 :C₆H₅CH₂N(CH₃)₂RCl (R=C₈H₁₇ to C₁₈H₃₇)
 :MFCD00145757