


Benzalkonium chloride 50% (w/w) in aqueous solution (alkyl distribution C8-C16)
ACRO263820010
:
- Pack type:Glass bottle
- Pk:1 L
- Seq:5037
- Appearance:Clear colorless to light yellow Solution
- Titration Argentometric:49 to 52 % (based on M.W.= 352.5)
- pH:6 to 9 (10 % in water)
- Impurity:≤2 % Free amine + amine HCl (M.W.= 226/262.5)
:
:0.98 g/cm³
:105 °C
:Ambient
:264-151-6
:1760
:8,II
:GHS07
:GHS05
:GHS09
:8001-54-5
:C₆H₅CH₂N(CH₃)₂RCl (R=C₈H₁₇ to C₁₈H₃₇)
:MFCD00145757