Tween® 20 (Polysorbate) 50% in aqueous solution
Catalog # INVT003005
Supplier: INVITROGEN
CAS Number:
Specifications
- Pk:1 KIT
- Seq:0075
- Environmentally Preferable:
Specifications
About this item
Synonyms: Polyoxyethylene sorbitan monolaurate
:
:1227.53 g/mol
Boiling Pt:100 °C (1013 hPa)
Density:1.1 g/cm³ (20 °C)
Flash Pt:>149 °C
Storage Temperature:Ambient
EINECS:500-018-3
CAS Number:9005-64-5
Formula:C₅₈H₁₁₄O₂₆
MDL Number:MFCD00165986