Tween® 20 (Polysorbate) 10% in aqueous solution
Catalog # BRAD161-0781
Supplier: BIO RAD
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:1 L
- Seq:5037
- Environmentally Preferable:
Specifications
About this item
Synonyms: Polyoxyethylene sorbitan monolaurate
:
:1227.53 g/mol
Storage Temperature:Ambient
EINECS:500-018-3
CAS Number:9005-64-5
Formula:C₅₈H₁₁₄O₂₆
MDL Number:MFCD00165986