
TRITON™ X-100 (Polyethylene glycol tert-octylphenyl ether) 10% in water, Sigma-Aldrich®
SIAL93443-100ML
:
- Pack type:Glass bottle
- Pk:100 mL
- Seq:2440
:
:∼100 °C (1013 hPa)
:∼1.01 g/cm³ (20 °C)
:GHS05
:Ambient
:9002-93-1
:9036-19-5
:MFCD00132505