Order Entry
China
ContactUsLinkComponent
Sucrose, EMPROVE® ESSENTIAL Ph. Eur., BP, NF, JP, SAFC®
Sucrose, EMPROVE® ESSENTIAL Ph. Eur., BP, NF, JP, SAFC®
Catalog # MERP1.07653.9029
CAS Number:  
undefined
Sucrose, EMPROVE® ESSENTIAL Ph. Eur., BP, NF, JP, SAFC®
Catalog # MERP1.07653.9029
CAS Number:  

Specifications

  • Pk:
    25 kg
  • Seq:
    4283

Specifications

About this item

 :
Merck Index:13,08966
Melting Pt:169…170 °C
Density:1.29 g/cm³ (20 °C)
 :342.3 g/mol
Storage Temperature:Ambient
EINECS:200-334-9
CAS Number:57-50-1
Formula:C₁₂H₂₂O₁₁
MDL Number:MFCD00006626