

Chloroform-[D₁] 99.8 atom % D + 1% TMS stabilised for NMR spectroscopy
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:865-49-6
:
Formula:CDCl₃
MDL Number:MFCD00000827
:120.38 g/mol
Boiling Pt:60.8 °C (1013 hPa)
Melting Pt:–64 °C
Density:1.5 g/cm³ (20 °C)
Storage Temperature:Refrigerator
EINECS:212-742-4
ADR:6.1,III
:Stabilised with silver
UN:1888
:GHS06
:GHS08
Specification Test Results
- Appearance:Clear colorless Liquid
- Water:≤0.02 %
- Stabilizer:silver foil
- D-Enrichment:≥99.75 %
- TMS:1 v/v%