Order Entry
China
ContactUsLinkComponent
VWR® Talon®, Support stand with rod and double burette clamp
VWR® Talon®, Support stand with rod and double burette clamp
Catalog # VWRU89202-636
undefined
VWR® Talon®, Support stand with rod and double burette clamp
Catalog # VWRU89202-636
Specifications
  • Description:
    Double burette clamp and support stand with rod, complete
  • W×D×H:
    178×330×25* mm
  • Pk:
    1
Specifications
About this item
This stand with double burette clamp holds any size burette from micro to 100 ml capacity.
  • Electropolished stainless steel finish
  • Graduated markings
  • Easily adjusted height
  • Stainless steel support rod
Simply compress the scissor-like mechanism, insert burette, and gently release to grip. Numbers and graduation on burette remain easy to read. For height adjustments, recompress mechanism and slide burette up or down and gently release. Clamp unit attaches to optional standard support rod with built-in reinforced hook connector. Adjusting nut faces forward for easy use. The stainless steel rod attaches to the optional porcelain base.
Support rod: 13×578 mm
Caution: * Porcelain base