Vydac 302IC, anion exclusion HPLC columns
: HICHROM
Vydac 302IC is a low capacity anion-exchange (quaternary amine) material based on a high purity 10 µm large pore size silica.
- Well referenced for 'non-suppressed' ion chromatography applications
- Used with standard HPLC systems for analysis of common ions
- Widely used for environmental analysis
- Catalog No:
- HICH302IC4.6
- HICH302GD4/N
- Environmentally Preferable:
- Low Manufacturing Impact
- Low Manufacturing Impact
- Environmental certification:
- ISO 14001:2015 (530840 UM15)
- ISO 14001:2015 (530840 UM15)