Order Entry
China
ContactUsLinkComponent
VWR®, Peristaltic Tubing Pump, Variable-Speed
VWR®, Peristaltic Tubing Pump, Variable-Speed
Certificates
undefined
VWR®, Peristaltic Tubing Pump, Variable-Speed
Specifications
  • Catalog No:
  • VWRU70730-064
  • VWRU70730-062
  • Model:
  • Low flow
  • Medium flow
  • Min. flow rate:
  • 0,03 ml/min
  • 0,4 ml/min
  • Max. flow rate:
  • 8,2 ml/min
  • 85 ml/min
  • W×D×H:
  • 168,3×120,6×114,3 mm
  • 168,3×120,6×114,3 mm
  • Weight:
  • 0,56 kg
  • 0,56 kg
Specifications