Order Entry
China
ContactUsLinkComponent
Magnetic stirrer heads with conical joint, for vaccum
  JUCH10220.00
undefined
Magnetic stirrer heads with conical joint, for vaccum
  JUCH10220.00
  • Description:
    Stirrer head, cone B 29
  • Seq:
    01
  • Pk:
    1
Conical joint stirrer heads which have a stainless steel bottom, special steel shaft and chromium-plated brass upper body for vacuum.