VWR® sympHony™, Multi-parameter meters, handheld
Specially designed for use in environmental field applications and in the lab, these rugged and durable handheld meters are used with probes to measure various parameters in solutions.
- Simple to operate, intuitive user interface with step-by-step on screen prompts
- Consistently accurate and reliable measurements
- Configurable to each user's unique needs
- Waterproof casing (IP67 certified)
Each meter features one single connector that can be used when testing for multiple parameters, and also allows for automatic or manual calibration and automatic buffer recognition.
: Calibrated pH meters and kits include a Troemner ISO/IEC Accredited Calibration Certificate traceable to NIST. The ISO/IEC 17025 Accredited Calibration Certificate includes electrical simulation of multiple data points within the functional ranges with the associated uncertainties. Troemner is accredited by NVLAP under Laboratory Code 105013-0 to perform ISO/IEC 17025 calibrations.
: The meters are available without electrodes or as kits including electrodes.
- Catalog No:
- VWRU89231-668
- VWRU89231-688
- Model:
- VWR sympHony™
- pH range:
- −2...+19
- pH resolution:
- 0,1/0,01/0,001 (selectable)
- pH accuracy:
- ≤ 0,01 (± 1 digit)
- pH calibration:
- Up to 5 points
- mV range:
- ± 2000
- mV resolution:
- 0,1/1
- mV accuracy:
- ≤ 0,2 (± 1 digit)
- Conductivity range:
- 0,001 µS/cm - 1000 mS/cm
- Conductivity resolution:
- Variable (autorange)
- Conductivity accuracy:
- ≤ 0,5 % of the measured value (± 1 digit)
- Conductivity calibration:
- Up to 3 points
- Resistivity range:
- 1 Ω - 1000 MΩ
- Resistivity accuracy:
- ≤0,5% (±1 digit)
- Salinity range:
- 0,0 - 42 ppt
- TDS range:
- 0,000 mg/l - 4444,4 g/l
- DO concentration:
- 0,00 - 60,0 mg/L
- DO concentration resolution:
- 0,01 mg/L (0,00 - 19,99 mg/L); 0,1 mg/L (20,0 - 60,0 mg/L)
- DO saturation:
- 0,0 - 600 %
- DO accuracy:
- ≤ 0,5 % of the measured value (± 1 digit)
- DO barometric pressure range:
- 600 - 1133 mBar
- Temperature range:
- −20...+150 °C
- Temperature resolution:
- 0,1 °C
- Temperature accuracy:
- ≤ 0,2 (± 1 digit)
- Temperature compensation:
- Via an ATC probe or manual entry
- Power supply:
- 4 AA alkaline batteries or nickel metal hydride (NiMH) rechargeable batteries
- Compliance:
- IP 67
- Data logging capacity:
- Up to 500 measurements
- Connections:
- MP-5 connector (IP 67) for models H10P, H10C and H10D
MP-8 connector (IP 67) for models H30PCO and H30PCD
- W×D×H:
- 194×92×43 mm
- Weight:
- 0,4 kg