Order Entry
China
Orders LinkContactUsLinkComponent
Endotoxin, Reference Standard

Endotoxin, Reference Standard

Endotoxin, USP reference standard, 11,000 USP Endotoxin Units, Vial, 1
Catalog # USPH1235503
Supplier:   U.S. PHARMACOPEIA
Endotoxin, Reference Standard
Catalog # USPH1235503
Supplier:  U.S. PHARMACOPEIA
CAS Number:  
Endotoxin, Reference Standard

Endotoxin, USP reference standard, 11,000 USP Endotoxin Units, Vial, 1

CAS Number:  
Price
Inquire For Price
Log in for availability
1 * 1 件
Product Details & Documents

USP Reference Standards are specified for use in conducting official USP–NF tests and assays. USP also provides Reference Standards specified in the Food Chemicals Codex as well as authentic substances—high-quality chemical samples—as a service to analytical, clinical, pharmaceutical and research laboratories. To confirm accuracy and reproducibility, USP Reference Standards are rigorously tested and evaluated by multiple independent laboratories including USP, commercial, regulatory, and academic labs. USP also provide publicly available, official documentary standards for pharmaceutical ingredients in the USP–NF that link directly with our primary reference standards.

Certifications: US Pharmacopea Reference Standard

Caution: Cold Shipment required

Specifications

Specifications

  • Pack type:
    Vial
  • Pk:
    1
  • Standards:
    USP reference standard
  • Description:
    Endotoxin
  • Pk Info:
    11,000 USP Endotoxin Units

Specifications