Organic reference standard, Povidone, Ph.Eur., RS
Catalog # EDQMP2660000
Supplier: EDQM
CAS Number:
Specifications
- Pack type:Vial
- Pk:50 mg
- Standards:Reference standard
- CAS No.:9003-39-8
- Description:Povidone
- Formula:C₆H₉NO
- Storage temperature:Ambient
- Description:Povidone
Specifications
About this item
- In Monograph 0685
- Solubility in water: ≥ 270 g/l at 23 °C