VWR® Signature™, Single channel pipettes, high-performance, variable volume
Supplier: VWR BRAND PRIVATE LABEL USA
Certificates
About this item
This pipettes features an ergonomic design for comfortable pipetting, superior accuracy and precision. Ideal for critical applications.
- Extended, curved finger rest allows the pipette to balance on the index finger
- Both plunger and volume-adjustment forces have been significantly reduced to eliminate the strain of repetitive pipetting
- Volume setting is continuously adjustable using either the micrometer wheel or dispenser button
- Locking mechanism prevents accidental volume shift while the instrument is in use
- Easy-grip ejector system provides adjustment of the ejector height
- The stainless steel construction of the ejector prevents corrosion and bending
- Fully autoclavable
Delivery information: Supplied with a tool for in-lab calibration and a three-year warranty.