Order Entry
China
ContactUsLinkComponent
Retinyl Acetate, Reference Standard
Retinyl Acetate, Reference Standard
Catalog # USPH1716002
Supplier:  U.S. PHARMACOPEIA
CAS Number:  
Retinyl Acetate, Reference Standard
Catalog # USPH1716002
Supplier:  U.S. PHARMACOPEIA
CAS Number:  

Specifications

  • Pk:
    5
  • Standards:
    USP reference standard
  • CAS No.:
    127-47-9
  • Description:
    Retinyl Acetate
  • Synonym(s):
    Vitamin A
  • Formula:
    C₂₂H₃₂O₂
  • Description:
    Retinyl Acetate
  • Pk Info:
    10 ampules x 0.5 g

Specifications

About this item

USP Reference Standards are specified for use in conducting official USP–NF tests and assays. USP also provides Reference Standards specified in the Food Chemicals Codex as well as authentic substances—high-quality chemical samples—as a service to analytical, clinical, pharmaceutical and research laboratories. To confirm accuracy and reproducibility, USP Reference Standards are rigorously tested and evaluated by multiple independent laboratories including USP, commercial, regulatory, and academic labs. USP also provide publicly available, official documentary standards for pharmaceutical ingredients in the USP–NF that link directly with our primary reference standards.

Certifications: US Pharmacopea Reference Standard