VWR®, Microcentrifuge tubes, with screw cap, standard line
Catalog # VWRU10025-756
Supplier: VWR BRAND PRIVATE LABEL USA
Specifications
- Type:without screw cap
- 版本:Free standing, sterile
- 容量:2 ml
- 顺序:03
- 包装规格:500
- 最大相对离心力:20000×g
Specifications
About this item
These are crystal-clear PP tubes that are available in conical bottom and self-standing bottom formats and ideal for sample processing applications.
- Tubes can be spun up to 20000 ×g
- Autoclavable and freezable
- DNase-free, RNase-free and non-pyrogenic
Certifications: TSE/BSE-free certification.
Ordering information: Caps and color inserts are has to be ordered seperately.
Packaging: Bagged tubes are packaged in durable and re-sealable (zip closure) bags. Sterile tubes are sold with attached cap and non sterile tubes are sold without cap.