Labels, Write-On NFPA
Catalog # NAMAHMS12L
Supplier: NATIONAL MARKER
Specifications
- Colour:Multicolour
- Description:Write-on NFPA labels
- Dimensions:51×51 mm
- Material:Paper
- Pk:250
Specifications
About this item
The legend for this hazardous materials label reads health hazard, fire hazard, specific hazard, and instability.
- Pressure-sensitive vinyl with pressure-sensitive adhesive
- Waterproof and graffiti-proof
- Resistant to chemicals
Colour: Multicolour
Certifications: National Fire Protection Association (NFPA)
Packaging: Roll-type.