Order Entry
China
Orders LinkContactUsLinkComponent
 

Trypsin

5×20 g, 20 µG
  PROMV5111
 :   PROMEGA
Trypsin
  PROMV5111
 :  PROMEGA
 :  
Trypsin
Trypsin

5×20 g, 20 µG

 :  
 
 
   
1 * 20 µG
 
   
 

Trypsin is a serine protease that specifically cleaves at the carboxylic side of lysine and arginine residues. The stringent specificity of trypsin is essential for protein identification.

  • Lysine residues modified by reductive methylation, yielding a highly active and stable trypsin
  • Lyophilised

The specificity of the purified trypsin is further improved by TPCK treatment, which inactivates chymotrypsin. The treated trypsin is then purified by affinity chromatography and lyophilised.

It is resistant to mild denaturing conditions such as 0,1% SDS, 1 M urea or 10% acetonitrile and retains 50% of its activity in 2 M guanidine HCl. The activity of trypsin is decreased when acidic residues are present on either side of a susceptible bond. If proline is at the carboxylic side of lysine or arginine, the bond is almost completely resistant to cleavage.

Reaction buffer: 50 mM NH₄HCO₃ (pH 7,8).

 : Kit includes Sequencing grade modified trypsin, porcine, frozen and trypsin resuspension dilution buffer.

 

 

 

  • Pk:
    20 µG
  • Pk Info:
    5×20 g
  • Cat. No.:
    PROMV5111
  • Form:
    Lyophilised
  • Storage conditions:
    –30...–10 °C
  • Grade:
    Sequencing grade
  • Enzyme name:
    Trypsin

 

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment.