Order Entry
China
ContactUsLinkComponent
CytoTox 96® Non-Radioactive Cytotoxicity Assay
CytoTox 96® Non-Radioactive Cytotoxicity Assay
Catalog # PROMG1780
Supplier:  PROMEGA
CAS Number:  
undefined
CytoTox 96® Non-Radioactive Cytotoxicity Assay
Catalog # PROMG1780
Supplier:  PROMEGA
CAS Number:  
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 
Specifications
  • Pk:
    1.000 Assays
  • Description:
    CytoTox 96® Non-Radioactive Cytotoxicity Assay
Specifications
About this item
The CytoTox 96® Non-Radioactive Cytotoxicity Assay quantitatively measures lactate dehydrogenase (LDH), a stable cytosolic enzyme that is released upon cell lysis, in much the same way as [⁵¹Cr] is released in radioactive assays.
  • Non-radioactive, time-saving assay
  • Highly sensitive assay
  • Low-level damage to cell membranes
Released LDH in culture supernatants is measured with a 30-minute coupled enzymatic assay that results in the conversion of a tetrazolium salt (INT) into a red formazan product. The amount of colour formed is proportional to the number of lysed cells. Visible wavelength absorbance data are collected using a standard 96-well plate reader.
The CytoTox 96® Non-Radioactive Cytotoxicity Assay requires no radioactive waste disposal or [⁵¹Cr] and eliminates labeling of target cells prior to conducting experiments. Standard equipment can be used to collect absorbance (visible wavelength) data with a standard 96-well plate reader.
The assay can be used to measure membrane integrity for cell-mediated cytotoxicity assays in which a target cell is lysed by an effector cell, or to measure lysis of target cells by bacteria, viruses, proteins, chemicals, etc.
Delivery: Kit includes Substrate Mix, Assay Buffer, LDH positive control, Lysis Solution (10X) and Stop Solution.