Order Entry
China
Orders LinkContactUsLinkComponent
Sterilisation indicators

Sterilisation indicators

Self-adhesive crepe paper with thermo-active pigments, 50 m, 19 mm
Catalog # BRND61750
Supplier:   BRAND
Sterilisation indicators
Catalog # BRND61750
Supplier:  BRAND
Sterilisation indicators
Sterilisation indicators

Self-adhesive crepe paper with thermo-active pigments, 50 m, 19 mm

Price
Inquire For Price
Log in for availability
1 * 1 件
Product Details & Documents

This self-adhesive crepe indicator tape turns from white to brown upon autoclaving.

Specifications
  • Width:
    19 mm
  • Description:
    Self-adhesive crepe paper with thermo-active pigments
  • Length:
    50 m
  • Seq:
    01