Order Entry
China
ContactUsLinkComponent
Endoproteinase Asp-N, sequencing grade
Endoproteinase Asp-N, sequencing grade
Catalog # PROMV1621
Supplier:  PROMEGA
CAS Number:  
undefined
Endoproteinase Asp-N, sequencing grade
Catalog # PROMV1621
Supplier:  PROMEGA
CAS Number:  
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 
Specifications
  • Pk:
    2 µG
  • Description:
    Endoproteinase Asp-N, sequencing grade, Pseudomonas fragi
Specifications
About this item
Endoproteinase hydrolyses peptide bonds on the N-terminal side of aspartic and cysteic acid residues (Asp and Cys).
  • Optimal activity in pH range of 4 to 9
  • Sequencing grade enzyme for use alone or in combination with other proteases
  • Use for in-solution or in-gel digestion reactions
This enzyme can be used alone or in combination with other proteases to produce protein digests for peptide mapping applications or protein identification by peptide mass fingerprinting or MS/MS spectral matching. It is also suitable for digestion reactions in solution or in gel.