Order Entry
China
ContactUsLinkComponent
dNTP mix
dNTP mix
Catalog # PROMU1515
Supplier:  PROMEGA
CAS Number:  
undefined
dNTP mix
Catalog # PROMU1515
Supplier:  PROMEGA
CAS Number:  
Specifications
  • Pk:
    1.000 µl
  • Description:
    dNTP mix, 10 mM
  • Seq:
    02
Specifications
About this item
dNTP Mix is a solution containing sodium salts of dATP, dCTP, dGTP and dTTP, each at 10 mM in water at pH 7,5. The total concentration of nucleotides is 40 mM. One microliter of the dNTP Mix in a 50 μl reaction will give a final dNTP concentration of 200 μM for each dNTP.
  • Highly pure (>99% triphosphate)
  • 10 mM each nucleotide in water
  • Reduce pipetting steps and possibility of contamination by adding nucleotides as a mix