Avantor® Prevail C18-Select, HPLC Columns
Supplier: HICHROM
Certificates
About this item
Prevail™ HPLC columns exhibit long column lifetime in both highly aqueous and highly organic mobile phases. All phases, apart from Prevail™ Carbohydrate ES are silica-based. The stability of these phases is such that a single column can be switched between highly aqueous for analysis of highly polar analytes and highly organic for strong retention of hydrophobic analytes.
- Stable from highly organic to highly aqueous
- Speciality phases for specific applications
- Excellent sensitivity with microbore and ELSD applications
Same benefits as Prevail™ C18, but slightly more retentive.
Accessories information: Use guard cartridges with holder (80101/N) and column coupler (HI-081).
Specifications
- Catalog No:
- HICH99316
- HICH99309
- HICH96691/N
- HICH99119/N
- HICH99302
- HICH99300
- HICH99313
- HICH99301
- Environmentally Preferable:
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- USP code:
- L1
- L1
- L1
- L1
- L1
- L1
- Environmental certification:
- ISO 14001:2015 (530840 UM15)
- ISO 14001:2015 (530840 UM15)
- ISO 14001:2015 (530840 UM15)
- ISO 14001:2015 (530840 UM15)
- ISO 14001:2015 (530840 UM15)
- ISO 14001:2015 (530840 UM15)
- ISO 14001:2015 (530840 UM15)
- ISO 14001:2015 (530840 UM15)