pH indicator test strips, BDH®
About this item
These test trips are used for rapid pH testing directly at the point of interest. Suitable for careful testing of dangerous, poisonous and aggressive liquids as the long handle prevents fingers from coming into contact with the sample.
- Enables high precise pH determination
- Strips are available in 13 different pH ranges with accuracy down to 0,2 pH units
- The dye is chemically bond to the test strip to protect against contamination
- Colour chart consists of 4 different colour blocks for each pH value
Packaging: 100 strips of size 6×85 mm packaged in a flat plastic box.
2 Options Available Below
Product Details & Documents
These test trips are used for rapid pH testing directly at the point of interest. Suitable for careful testing of dangerous, poisonous and aggressive liquids as the long handle prevents fingers from coming into contact with the sample.
- Enables high precise pH determination
- Strips are available in 13 different pH ranges with accuracy down to 0,2 pH units
- The dye is chemically bond to the test strip to protect against contamination
- Colour chart consists of 4 different colour blocks for each pH value
Packaging: 100 strips of size 6×85 mm packaged in a flat plastic box.
Product Family Options
Product Information
- pH rangepH gradationAvailabilityPrice
- BDHB83930.601pH indicator strips
- BDHB35317.604pH indicator strips
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.



