Order Entry
China
Orders LinkContactUsLinkComponent
 

Buffers, pH reference standards, BDH®

Buffers, pH reference standards, BDH®
Supplier:  BDH US PRODUCED
Certificates
Buffers, pH reference standards, BDH®

18 Options Available Below

Product Details & Documents

Buffers are available as colourless and coloured solutions.

 : ISO 17025 and traceable to NIST.

Product Family Options

Product Information

  • pH value
    pH accuracy
    Colour
    Reference temperature
    Pack type
    Pk
    Availability
    Price
  • BDHBBDH5006-500ML
    Buffer, pH reference standard
    Buffer, pH reference standard
    1,68
    ±0,01
    Colourless
    25 °C
    Poly bottle
    500 mL
    Log in for availability
    Log in for availability
    1 * 500 毫升
    1 * 500 毫升

  • BDHB5010-500ML
    Buffer, pH reference standard
    Buffer, pH reference standard
    2,00
    ±0,01
    Colourless
    25 °C
    Poly bottle
    500 mL
    Log in for availability
    Log in for availability
    1 * 500 毫升
    1 * 500 毫升

  • BDHB5014-500ML
    Buffer, pH reference standard
    Buffer, pH reference standard
    3,00
    ±0,01
    Colourless
    25 °C
    Poly bottle
    500 mL
    Log in for availability
    Log in for availability
    1 * 500 毫升
    1 * 500 毫升

  • BDHB5018-500ML
    Buffer, pH reference standard
    Buffer, pH reference standard
    4,00
    ±0,01
    Red
    25 °C
    Poly bottle
    500 mL
    Log in for availability
    Log in for availability
    1 * 500 毫升
    1 * 500 毫升

  • BDHB5024-500ML
    Buffer, pH reference standard
    Buffer, pH reference standard
    4,00
    ±0,01
    Colourless
    25 °C
    Poly bottle
    500 mL
    Log in for availability
    Log in for availability
    1 * 500 毫升
    1 * 500 毫升

  • BDHB0198-20L
    Buffer, pH reference standard
    Buffer, pH reference standard
    4,00
    ±0,01
    Red
    25 °C
    Cubitainer®
    20 L
    Log in for availability
    Log in for availability
    1 * 20 升
    1 * 20 升

  • BDHB5034-500ML
    Buffer, pH reference standard
    Buffer, pH reference standard
    5,00
    ±0,01
    Colourless
    25 °C
    Poly bottle
    500 mL
    Log in for availability
    Log in for availability
    1 * 500 毫升
    1 * 500 毫升

  • BDHB5038-500ML
    Buffer, pH reference standard
    Buffer, pH reference standard
    6,00
    ±0,01
    Colourless
    25 °C
    Poly bottle
    500 mL
    Log in for availability
    Log in for availability
    1 * 500 毫升
    1 * 500 毫升

  • BDHB5040-500ML
    Buffer, pH reference standard
    Buffer, pH reference standard
    6,86
    ±0,01
    Colourless
    25 °C
    Poly bottle
    500 mL
    Log in for availability
    Log in for availability
    1 * 500 毫升
    1 * 500 毫升

  • BDHB5046-500ML
    Buffer, pH reference standard
    Buffer, pH reference standard
    7,00
    ±0,01
    Yellow
    25 °C
    Poly bottle
    500 mL
    Log in for availability
    Log in for availability
    1 * 500 毫升
    1 * 500 毫升

  • BDHB5052-500ML
    Buffer, pH reference standard
    Buffer, pH reference standard
    7,00
    ±0,01
    Colourless
    25 °C
    Poly bottle
    500 mL
    Log in for availability
    Log in for availability
    1 * 500 毫升
    1 * 500 毫升

  • BDHB0194-20L
    Buffer, pH reference standard
    Buffer, pH reference standard
    7,00
    ±0,01
    Yellow
    25 °C
    Cubitainer®
    20 L
    Log in for availability
    Log in for availability
    1 * 20 升
    1 * 20 升

  • BDHB5060-500ML
    Buffer, pH reference standard
    Buffer, pH reference standard
    8,00
    ±0,01
    Colourless
    25 °C
    Poly bottle
    500 mL
    Log in for availability
    Log in for availability
    1 * 500 毫升
    1 * 500 毫升

  • BDHB5062-500ML
    Buffer, pH reference standard
    Buffer, pH reference standard
    9,00
    ±0,01
    Colourless
    25 °C
    Poly bottle
    500 mL
    Log in for availability
    Log in for availability
    1 * 500 毫升
    1 * 500 毫升

  • BDHB5066-500ML
    Buffer, pH reference standard
    Buffer, pH reference standard
    9,18
    ±0,01
    Colourless
    25 °C
    Poly bottle
    500 mL
    Log in for availability
    Log in for availability
    1 * 500 毫升
    1 * 500 毫升

  • BDHB5072-500ML
    Buffer, pH reference standard
    Buffer, pH reference standard
    10,00
    ±0,01
    Blue
    25 °C
    Poly bottle
    500 mL
    Log in for availability
    Log in for availability
    1 * 500 毫升
    1 * 500 毫升

  • BDHB5078-500ML
    Buffer, pH reference standard
    Buffer, pH reference standard
    10,00
    ±0,01
    Colourless
    25 °C
    Poly bottle
    500 mL
    Log in for availability
    Log in for availability
    1 * 500 毫升
    1 * 500 毫升

  • BDHB5086-500ML
    Buffer, pH reference standard
    Buffer, pH reference standard
    12,00
    ±0,02
    Colourless
    25 °C
    Poly bottle
    500 mL
    Log in for availability
    Log in for availability
    1 * 500 毫升
    1 * 500 毫升