Test kits, photometric, Zinc, LCK and LCS cuvette tests
HACHLCK360
: Hach
:
- Chemical method:PAR
- Pk:24 Tests
- Description:Zinc
- Seq:295
- Type:LCK360
- For unit:DR3900, DR6000, DR1900, DR2800, DR3800, DR5000
- Range:0,2 - 6,0 mg/l Zn
Pre-dosed reagents for maximum safety.
- Precise and reliable measurement values
- Maximum safety for users, thanks to the closed system and low amounts of reagents
- Convenient and error-free dosing of the reagents
- Barcode label for automatic recognition in the photometer
- Differentiation between tests and measuring ranges by means of colour-coding
Storage conditions: 15 to 25 °C