Test kits, photometric, Water hardness, LCK cuvette tests
Catalog # HACHLCK327
Supplier: HACH
CAS Number:
Specifications
- Chemical method:Metalphthalein
- Pk:25 Tests
- Description:Hardness (Ca + Mg)
- Seq:236
- Type:LCK327
- For unit:DR3900, DR6000, DR1900, DR2800, DR3800, DR5000
- Range:1 - 20 °dH
5 - 100 mg/l Ca
Specifications
About this item
Pre-dosed reagents for maximum safety.
- Precise and reliable measurement values
- Maximum safety for users, thanks to the closed system and low amounts of reagents
- Convenient and error-free dosing of the reagents
- Barcode label for automatic recognition in the photometer
- Differentiation between tests and measuring ranges by means of colour-coding
Storage conditions: 15 to 25 °C