Order Entry
China
ContactUsLinkComponent
Test kits, photometric, Surfactants, anionic, LCK cuvette test with reagents
Test kits, photometric, Surfactants, anionic, LCK cuvette test with reagents
  HACHLCK332
 :  Hach
 :  
Test kits, photometric, Surfactants, anionic, LCK cuvette test with reagents
  HACHLCK332
 :  Hach
 :  
  • Chemical method:
    Methylene Blue (MBA)
  • Pk:
    25 Tests
  • Description:
    Surfactants (anionic)
  • Seq:
    273
  • Type:
    LCK332
  • For unit:
    DR3900, DR6000, DR1900, DR2800, DR3800, DR5000
  • Range:
    0,05 - 2,0 mg/l
Ready for use, photometric evaluation system and environmentally friendly.
  • Precise and reliable measurement values
  • Maximum safety for users, thanks to the closed system and low amounts of reagents
  • Convenient and error-free dosing of the reagents
  • Barcode label for automatic recognition in the photometer
  • Differentiation between tests and measuring ranges by means of colour-coding
Storage conditions: 15 to 25 °C
 : ISO 7875-1-2-1984, DIN 38409-H 23-1