Test kits, photometric, Nitrite, LCK cuvette tests
Catalog # HACHLCK341
Supplier: HACH
CAS Number:
Specifications
- Chemical method:Diazotisation
- Pk:25 Tests
- Description:Nitrite
- Seq:254
- Type:LCK341
- For unit:DR3900, DR6000, DR1900, DR2800, DR3800, DR5000
- Range:0,015 - 0,6 mg/l NO₂-N
0,05 - 2 mg/l NO₂
Specifications
About this item
Pre-dosed reagents for maximum safety.
- Precise and reliable measurement values
- Maximum safety for users, thanks to the closed system and low amounts of reagents
- Convenient and error-free dosing of the reagents
- Barcode label for automatic recognition in the photometer
- Differentiation between tests and measuring ranges by means of colour-coding
Storage conditions: 15 to 25 °C
Certifications: EN ISO 26777, DIN 38405 D10