Order Entry
China
ContactUsLinkComponent
Test kits, photometric, Chromium, LCK cuvette tests
Test kits, photometric, Chromium, LCK cuvette tests
Catalog # HACHLCK313
Supplier:  HACH
CAS Number:  
undefined
Test kits, photometric, Chromium, LCK cuvette tests
Catalog # HACHLCK313
Supplier:  HACH
CAS Number:  

Specifications

  • Chemical method:
    Diphenylcarbazide
  • Pk:
    25 Tests
  • Description:
    Chromium (total + VI)
  • Seq:
    215
  • Type:
    LCK313
  • For unit:
    DR3900, DR6000, DR1900, DR2800, DR3800, DR5000
  • Range:
    0,03 - 1,0 mg/l Cr VI
    0,03 - 1,0 mg/l Cr total

Specifications

About this item

Pre-dosed reagents for maximum safety.

  • Precise and reliable measurement values
  • Maximum safety for users, thanks to the closed system and low amounts of reagents
  • Convenient and error-free dosing of the reagents
  • Barcode label for automatic recognition in the photometer
  • Differentiation between tests and measuring ranges by means of colour-coding

Storage conditions: 2 to 8 °C (keep refrigerated)

Certifications: EN ISO 11083, DIN 38405-D24