Order Entry
China
ContactUsLinkComponent
Test kits, photometric, Chloride, LCK cuvette tests
Test kits, photometric, Chloride, LCK cuvette tests
Catalog # HACHLCK311
Supplier:  Hach
CAS Number:  
Test kits, photometric, Chloride, LCK cuvette tests
Catalog # HACHLCK311
Supplier:  Hach
CAS Number:  

Specifications

  • Chemical method:
    Iron (III) thiocyanate
  • Pk:
    24 Tests
  • Description:
    Chloride
  • Seq:
    211
  • Type:
    LCK311
  • For unit:
    DR3900, DR6000, DR1900, DR2800, DR3800, DR5000
  • Range:
    1 - 70 mg/l Cl
    70 - 1000 mg/l Cl

Specifications

About this item

Pre-dosed reagents for maximum safety.

  • Precise and reliable measurement values
  • Maximum safety for users, thanks to the closed system and low amounts of reagents
  • Convenient and error-free dosing of the reagents
  • Barcode label for automatic recognition in the photometer
  • Differentiation between tests and measuring ranges by means of colour-coding

Storage conditions: 2 to 8 °C (keep refrigerated)