Tetrachloroethylene, CMOS for microelectronic, J.T.Baker®
Catalog # BAKR9453-03
Supplier: Avantor
CAS Number:
Specifications
- Pk:4 L
- Seq:5162
- Environmentally Preferable:
Specifications
Specification Test Results
- Assay (Cl₂C:CCl₂) (by GC):≥ 99.0 %
- Color (APHA):≤ 10
- Residue after Evaporation:≤ 5 ppm
- Acidity (meq/g):≤ 0.0006
- Water (H₂O)(by Karl Fischer titrn):≤ 0.05 %
- Heavy Metals (as Pb):≤ 0.20 ppm
- Chloride (Cl):≤ 1.0 ppm
- Phosphate (PO₄):≤ 0.1 ppm
- Trace Impurities - Aluminum (Al):≤ 1 ppm
- Arsenic and Antimony (as As):≤ 0.010 ppm
- Trace Impurities - Barium (Ba):≤ 1.000 ppm
- Trace Impurities - Boron (B):≤ 0.010 ppm
- Trace Impurities - Cadmium (Cd):≤ 1.000 ppm
- Trace Impurities - Calcium (Ca):≤ 1 ppm
- Trace Impurities - Chromium (Cr):≤ 0.500 ppm
- Trace Impurities - Cobalt (Co):≤ 0.100 ppm
- Trace Impurities - Copper (Cu):≤ 0.010 ppm
- Trace Impurities - Gallium (Ga):≤ 0.050 ppm
- Trace Impurities - Germanium (Ge):≤ 1.000 ppm
- Trace Impurities - Gold (Au):≤ 0.500 ppm
- Trace Impurities - Iron (Fe):≤ 0.1 ppm
- Trace Impurities - Lithium (Li):≤ 1.000 ppm
- Trace Impurities - Magnesium (Mg):≤ 1.000 ppm
- Trace Impurities - Manganese (Mn):≤ 1 ppm
- Trace Impurities - Nickel (Ni):≤ 0.010 ppm
- Trace Impurities - Potassium (K):≤ 1.000 ppm
- Trace Impurities - Silicon (Si):≤ 1.000 ppm
- Trace Impurities - Silver (Ag):≤ 0.500 ppm
- Trace Impurities - Sodium (Na):≤ 1.000 ppm
- Trace Impurities - Strontium (Sr):≤ 1.000 ppm
- Trace Impurities - Tin (Sn):≤ 1.000 ppm
- Trace Impurities - Zinc (Zn):≤ 1.000 ppm
Specification Test Results
About this item
Synonyms: Tetrachloroethene, Ethylene tetrachloride, PCE, Perchloroethylene
:
:165.83 g/mol
ADR:6.1,III
:Management of Change (MOC) category = R
UN:1897
CAS Number:127-18-4
Formula:C₂Cl₄
MDL Number:MFCD00000834