
Sodium metabisulphite 65.0 - 67.4%, granular NF, FCC, Macron Fine Chemicals™
Supplier: MACRON AVANTOR BRAND
Certificates
About this item
CAS Number:7681-57-4
:
Formula:Na₂S₂O₅
MDL Number:MFCD00167602
:190.11 g/mol
Merck Index:13,08712
EINECS:231-673-0
:GHS05
Storage Temperature:Ambient
Specification Test Results
- NF - Assay (SO₂):65.0 - 67.4 %
- NF - Identification A:Passes Test
- NF - Identification B:Passes Test
- NF - Thiosulfate (S₂O₃):≤0.05 %
- NF - Iron (Fe):≤0.002 %
- NF - Chloride (Cl):≤0.05 %
- FCC - Assay (Na₂S₂O₅):96.5 - 100.0 %
- FCC - Identification A:Passes Test
- FCC - Identification B:Passes Test
- FCC - Iron (Fe):≤0.002 %
- FCC - Lead (Pb):≤2 mg/kg
- FCC - Selenium (Se):≤5 mg/kg
- Solubility (10 in 100):Passes Test