

di-Sodium tetraborate decahydrate 99.5-105.0%, crystals, BAKER ANALYZED® ACS, J.T.Baker®
Supplier: Avantor
Certificates
About this item
CAS Number:1303-96-4
:
Formula:Na₂B₄O₇·10H₂O
MDL Number:MFCD00149193
:381.37 g/mol
Merck Index:13,08662
Boiling Pt:320 °C (1013 hPa)
Melting Pt:75 °C
Density:1.73 g/cm³
Storage Temperature:Ambient
REACH:01-2119490790-32
:GHS08
:GHS07
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specification Test Results
- For Laboratory,Research,or Manufacturing Use:99.5 - 105.0%
- Assay (Na₂B₄O₇·10H₂O):99.5 - 105.0 %
- Insoluble Matter:≤0.005 %
- pH of 0.01M Solution at 25°C:9.15 - 9.20
- Chloride (Cl):≤0.001 %
- Phosphate (PO₄):≤0.001 %
- Sulfate (SO₄):≤0.005 %
- Calcium (Ca):≤0.005 %
- Heavy Metals (as Pb):≤0.001 %
- Trace Impurities - Iron (Fe):≤5 ppm