Sodium hydrogen carbonate 99.0-100.5% (by acidimetry on anhydrous substance), powder USP, FCC for haemodialysis, Macron Fine Chemicals™
Supplier: MACRON AVANTOR BRAND
Certificates
About this item
CAS Number:144-55-8
:
Formula:NaHCO₃
MDL Number:MFCD00003528
:84.01 g/mol
Merck Index:13,08655
Melting Pt:270 °C
Density:2.159 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:205-633-8
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specification Test Results
- USP - Assay (NaHCO₃) (dried basis)(by acidimetry):99.0 - 100.5 %
- USP - Aluminum (Al):≤2 ppm
- USP - Arsenic (As):≤2 ppm
- USP - Calcium (Ca):≤0.01 %
- USP - Carbonate (CO₃):≤0.23 %
- USP - Chloride (Cl):≤0.015 %
- USP - Copper (Cu):≤1 ppm
- USP - Identification A:Passes Test
- USP - Identification B:Passes Test
- USP - Insoluble Substances:Passes Test
- USP - Iron (Fe):≤5 ppm
- USP - Limit of Ammonia:≤20 ppm
- USP - Limit of Sulfur Compounds:≤0.015 %
- USP - Loss on Drying:≤0.25 %
- USP - Magnesium (Mg):≤0.004 %
- USP - Normal Carbonate:Passes Test
- USP - Limit of Organics:≤0.01 %
- FCC - Assay (NaHCO₃) (dried basis)(by acidimetry):99.0 - 100.5 %
- FCC - Ammonia (as NH₃):Passes Test
- FCC - Identification A:Passes Test
- FCC - Identification B:Passes Test
- FCC - Insoluble Substances:Passes Test
- FCC - Lead (Pb):≤2 mg/kg
- FCC - Loss on Drying:≤0.25 %